Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979177.1 | XP_027834978.1 | 3 - 4 | 963 | 116 | |
XM_027979178.1 | XP_027834979.1 | 4 - 5 | 3628 | 116 | ||
XM_027979179.1 | XP_027834980.1 | 4 - 5 | 1034 | 116 | ||
XM_027979180.1 | XP_027834981.1 | 4 - 5 | 1252 | 116 | ||
XM_027979181.1 | XP_027834982.1 | 3 - 4 | 966 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |