Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020187.4 | XP_004020236.1 | 3 - 4 | 476 | 81 | |
XM_027960171.1 | XP_027815972.1 | 2 - 3 | 333 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |