Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961675.1 | XP_027817476.1 | 2 | 323 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |