Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972295.1 | XP_027828096.1 | 1 - 2 | 307 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |