Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098906.2 | XP_014954392.2 | 9 - 10 | 1219 | 84 | |
XM_027974287.1 | XP_027830088.1 | 9 - 10 | 1427 | 84 | ||
XM_027974288.1 | XP_027830089.1 | 9 - 10 | 1189 | 84 | ||
XM_027974289.1 | XP_027830090.1 | 8 - 9 | 1142 | 84 |