Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082647.2 | NP_001076116.1 | 5 - 6 | 570 | 77 | |
XM_008268046.2 | XP_008266268.1 | 4 - 5 | 576 | 77 | ||
GenBank mRNA | AB050592.1 | - | 5 - 6 | 572 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |