Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101722.1 | NP_001095192.1 | 5 - 6 | 594 | 66 | |
GenBank mRNA | X61639.1 | - | 594 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |