Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115764.4 | XP_001115764.2 | 1761 | 95 | ||
XM_028838834.1 | XP_028694667.1 | 1177 | 95 | |||
GenBank mRNA | JU484073.1 | - | 764 | 95 | ||
JV647721.1 | - | 603 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |