Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260665.1 | NP_001247594.1 | 124 | 91 | ||
XM_028827204.1 | XP_028683037.1 | 169 | 91 | |||
GenBank mRNA | JU320037.1 | - | 148 | 91 | ||
JU471482.1 | - | 124 | 91 | |||
JV531182.1 | - | 154 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |