Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117025.4 | XP_001117025.1 | 488 | 81 | ||
XM_002798668.3 | XP_002798714.1 | 488 | 81 | |||
GenBank mRNA | JV662460.1 | - | 119 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |