Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114606.4 | XP_001114606.1 | 747 | 76 | ||
GenBank mRNA | JU326548.1 | - | 503 | 76 | ||
JU326549.1 | - | 503 | 76 | |||
JV723274.1 | - | 476 | 76 |