Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193817.3 | NP_001180746.2 | 474 | 67 | ||
GenBank mRNA | JU322247.1 | - | 428 | 67 | ||
JU476528.1 | - | 425 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |