Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193988.2 | NP_001180917.1 | 1268 | 65 | ||
GenBank mRNA | JU328188.1 | - | 897 | 65 | ||
JV654683.1 | - | 1686 | 65 | |||
JV667254.1 | - | 1694 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |