Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195448.1 | NP_001182377.1 | 105 | |||
GenBank mRNA | AY680454.1 | - | 439 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |