Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145294.2 | XP_015000780.2 | 1209 | 79 | ||
XM_028852629.1 | XP_028708462.1 | 2674 | 79 | |||
XM_028852630.1 | XP_028708463.1 | 2653 | 79 | |||
GenBank mRNA | JU493760.1 | - | 2643 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |