Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266030.1 | NP_001252959.1 | 369 | 73 | ||
XM_015146115.2 | XP_015001601.1 | 349 | 73 | |||
XM_028852248.1 | XP_028708081.1 | 531 | 73 | |||
XM_028852249.1 | XP_028708082.1 | 478 | 73 | |||
GenBank mRNA | JU322432.1 | - | 369 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |