Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040415.2 | NP_001035505.1 | 654 | 78 | ||
GenBank mRNA | AY913830.1 | - | 656 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |