Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260794.1 | NP_001247723.1 | 533 | 96 | ||
GenBank mRNA | JU327154.1 | - | 524 | 96 | ||
JU327155.1 | - | 524 | 96 | |||
JV045210.1 | - | 533 | 96 | |||
JV600682.1 | - | 659 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |