Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260685.1 | NP_001247614.1 | 313 | 133 | ||
XM_015135932.2 | XP_014991418.1 | 333 | 133 | |||
GenBank mRNA | JU320755.1 | - | 315 | 133 | ||
JU472045.1 | - | 312 | 133 | |||
JV045316.1 | - | 313 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |