Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC715304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105454.4 | XP_001105454.2 | 284 | 133 | ||
XM_015135991.2 | XP_014991477.1 | 409 | 133 | |||
XM_028847719.1 | XP_028703552.1 | 391 | 133 | |||
GenBank mRNA | KU253645.1 | - | 87 | 133 | ||
KU253647.1 | - | 87 | 133 | |||
KU253648.1 | - | 87 | 133 | |||
KU253649.1 | - | 87 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |