Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260836.2 | NP_001247765.1 | 868 | 139 | ||
GenBank mRNA | JV048492.1 | - | 868 | 139 | ||
JV636423.1 | - | 848 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |