Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087330.4 | XP_001087330.1 | 627 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |