Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNSTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141933.2 | XP_014997419.2 | 603 | 135 | ||
XM_028850740.1 | XP_028706573.1 | 605 | 135 | |||
XM_028850741.1 | XP_028706574.1 | 607 | 135 | |||
GenBank mRNA | JU495945.1 | - | 333 | 135 | ||
JV075023.1 | - | 337 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |