Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Rh02853755_m1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Species:
Rhesus monkey
Assay Type:
Non-coding
Assay ID
Rh02853755_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Macaca mulatta
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9zxgn:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0