Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258126.1 | NP_001245055.1 | 311 | 140 | ||
GenBank mRNA | JU330972.1 | - | 187 | 140 | ||
JU474756.1 | - | 311 | 140 | |||
JV045203.1 | - | 325 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |