Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261140.1 | NP_001248069.1 | 330 | 90 | ||
GenBank mRNA | JU336535.1 | - | 330 | 90 | ||
JV594865.1 | - | 358 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |