Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195660.1 | NP_001182589.1 | 346 | 144 | ||
GenBank mRNA | CB308818.1 | - | 349 | 144 | ||
CO644659.1 | - | 332 | 144 | |||
DQ147924.1 | - | 249 | 144 | |||
JU326829.1 | - | 224 | 144 | |||
JU472778.1 | - | 334 | 144 | |||
JV045909.1 | - | 350 | 144 | |||
JV635773.1 | - | 358 | 144 |
Set Membership: |
![]() ![]() |