Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266945.1 | NP_001253874.1 | 357 | 128 | ||
XM_015150401.2 | XP_015005887.1 | 470 | 128 | |||
GenBank mRNA | JU320969.1 | - | 357 | 128 | ||
JU472380.1 | - | 355 | 128 | |||
JV044554.1 | - | 357 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |