Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261722.1 | NP_001248651.1 | 1086 | 151 | ||
XM_015128177.2 | XP_014983663.1 | 1418 | 151 | |||
XM_028841795.1 | XP_028697628.1 | 1381 | 151 | |||
GenBank mRNA | JU472622.1 | - | 1086 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |