Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261698.1 | NP_001248627.1 | 1466 | 104 | ||
XM_028846745.1 | XP_028702578.1 | 988 | 104 | |||
GenBank mRNA | JU335189.1 | - | 1463 | 104 | ||
JU335190.1 | - | 1463 | 104 | |||
JU335191.1 | - | 1463 | 104 | |||
JU335192.1 | - | 1463 | 104 | |||
JU335193.1 | - | 1463 | 104 | |||
JU335194.1 | - | 1463 | 104 | |||
JU335195.1 | - | 1463 | 104 | |||
JU476173.1 | - | 1466 | 104 | |||
JU476174.1 | - | 1466 | 104 | |||
JU476175.1 | - | 1466 | 104 | |||
JV717814.1 | - | 1457 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |