Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101213.4 | XP_001101213.2 | 618 | 135 | ||
XM_015121789.2 | XP_014977275.2 | 618 | 135 | |||
XM_015121790.2 | XP_014977276.2 | 549 | 135 | |||
XM_015121791.2 | XP_014977277.2 | 706 | 135 | |||
XM_015121792.2 | XP_014977278.2 | 558 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |