Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194544.3 | NP_001181473.2 | 1252 | 75 | ||
GenBank mRNA | JU336695.1 | - | 1254 | 75 | ||
JV602355.1 | - | 1256 | 75 | |||
JV725012.1 | - | 1246 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |