Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260889.1 | NP_001247818.1 | 526 | 97 | ||
GenBank mRNA | JU321059.1 | - | 545 | 97 | ||
JU473756.1 | - | 527 | 97 | |||
JU473757.1 | - | 527 | 97 | |||
JV043413.1 | - | 526 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |