Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265982.1 | NP_001252911.1 | 344 | 83 | ||
XM_015149228.2 | XP_015004714.1 | 372 | 83 | |||
GenBank mRNA | JU320779.1 | - | 341 | 83 | ||
JU494268.1 | - | 126 | 83 | |||
JV045472.1 | - | 344 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |