Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277450.1 | NP_001264379.1 | 2 - 3 | 442 | 79 | |
NM_019191.2 | NP_062064.1 | 2 - 3 | 470 | 79 | ||
GenBank mRNA | AB010147.1 | - | 1 - 2 | 244 | 79 | |
AB017912.1 | - | 2 - 3 | 336 | 79 | ||
AF056001.1 | - | 2 - 3 | 370 | 79 | ||
BC127497.1 | - | 2 - 3 | 314 | 8364718 | 79 | |
FQ230791.1 | - | 2 - 3 | 308 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |