Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106449.2 | NP_001099919.1 | 7 - 8 | 896 | 68 | |
NM_001270650.1 | NP_001257579.1 | 8 - 9 | 970 | 68 | ||
NM_001270651.1 | NP_001257580.1 | 8 - 9 | 943 | 68 | ||
NM_001270652.1 | NP_001257581.1 | 7 - 8 | 873 | 68 | ||
NM_001270653.1 | NP_001257582.1 | 7 - 8 | 900 | 68 | ||
XM_006232855.2 | XP_006232917.1 | 8 - 9 | 965 | 68 | ||
XM_008761282.1 | XP_008759504.1 | 7 - 8 | 931 | 68 | ||
GenBank mRNA | FM111895.1 | - | 4 - 5 | 352 | 68 | |
FQ214903.1 | - | 7 - 8 | 833 | 68 | ||
FQ215988.1 | - | 8 - 9 | 899 | 68 | ||
FQ216125.1 | - | 7 - 8 | 822 | 68 | ||
FQ216236.1 | - | 5 - 6 | 567 | 68 | ||
FQ216534.1 | - | 7 - 8 | 799 | 68 | ||
FQ217203.1 | - | 7 - 8 | 820 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |