Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022389.2 | NP_071784.1 | 5 - 6 | 847 | 63 | |
XM_006230892.2 | XP_006230954.1 | 4 - 5 | 595 | 63 | ||
XM_006230893.2 | XP_006230955.1 | 4 - 5 | 474 | 63 | ||
XM_006230894.2 | XP_006230956.1 | 5 - 6 | 494 | 63 | ||
XM_008760170.1 | XP_008758392.1 | 5 - 6 | 1581 | 63 | ||
XM_008760171.1 | XP_008758393.1 | 4 - 5 | 1242 | 63 | ||
XM_008760172.1 | XP_008758394.1 | 5 - 6 | 1141 | 63 | ||
XM_008760173.1 | XP_008758395.1 | 4 - 5 | 1121 | 63 | ||
GenBank mRNA | AB016800.1 | - | 3 - 4 | 538 | 63 | |
AF071500.1 | - | 4 - 5 | 450 | 63 | ||
AF272393.1 | - | 5 - 6 | 847 | 63 | ||
BC081688.1 | - | 3 - 4 | 538 | 7109221 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |