Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053584.2 | NP_446036.1 | 5 - 6 | 442 | 108 | |
XM_006246914.1 | XP_006246976.1 | 5 - 6 | 510 | 108 | ||
XM_008768132.1 | XP_008766354.1 | 4 - 5 | 313 | 108 | ||
GenBank mRNA | BC126068.1 | - | 5 - 6 | 446 | 7387185 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |