Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133401.1 | NP_596892.1 | 1 - 2 | 82 | 67 | |
XM_006235994.2 | XP_006236056.1 | 3 - 4 | 907 | 67 | ||
GenBank mRNA | AF257746.1 | - | 1 - 2 | 82 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |