Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF063102.1 | - | 6 - 7 | 1525 | 89 | |
AF081148.1 | - | 7 - 8 | 1971 | 89 | ||
AF081149.1 | - | 7 - 8 | 1971 | 89 | ||
AF081150.1 | - | 7 - 8 | 1971 | 89 | ||
AF081151.1 | - | 7 - 8 | 1971 | 89 | ||
AF081152.1 | - | 7 - 8 | 1971 | 89 | ||
AF081153.1 | - | 7 - 8 | 1971 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |