Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138827.1 | NP_620182.1 | 7 - 8 | 1181 | 86 | |
GenBank mRNA | BC061873.1 | - | 7 - 8 | 1109 | 5598829 | 86 |
M13979.1 | - | 7 - 8 | 1181 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |