Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271045.1 | NP_001257974.1 | 3 - 4 | 584 | 86 | |
NM_001271046.2 | NP_001257975.1 | 3 - 4 | 451 | 86 | ||
NM_017288.2 | NP_058984.1 | 3 - 4 | 665 | 86 | ||
GenBank mRNA | BC094523.1 | - | 3 - 4 | 584 | 7316686 | 86 |
FQ215379.1 | - | 1 - 2 | 242 | 86 | ||
FQ216374.1 | - | 3 - 4 | 451 | 86 | ||
M91808.1 | - | 3 - 4 | 664 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Rattus norvegicus Probe spans exons |