Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039035.2 | NP_001034124.2 | 6 - 7 | 725 | 86 | |
XM_006239642.1 | XP_006239704.1 | 6 - 7 | 756 | 86 | ||
XM_006239643.1 | XP_006239705.1 | 6 - 7 | 732 | 86 | ||
GenBank mRNA | FM079490.1 | - | 1 - 2 | 122 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |