Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008335.1 | NP_001008336.1 | 1 - 2 | 54 | 77 | |
GenBank mRNA | BC085859.1 | - | 1 - 2 | 54 | 7190333 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |