Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010920.1 | NP_001010920.1 | 3 - 4 | 299 | 71 | |
XM_006254998.2 | XP_006255060.1 | 2 - 3 | 345 | 71 | ||
XM_006254999.2 | XP_006255061.1 | 3 - 4 | 339 | 71 | ||
GenBank mRNA | AY569013.1 | - | 3 - 4 | 299 | 71 | |
BC095904.1 | - | 2 - 3 | 272 | 7110846 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |