Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145828.1 | NP_001139300.1 | 1 | 4155 | 115 | |
GenBank mRNA | FJ750588.1 | - | 1 | 3188 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |