Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250882.2 | XP_006250944.1 | 14 - 15 | 1429 | 105 | |
XM_006250884.2 | XP_006250946.1 | 13 - 14 | 1358 | 105 | ||
XM_006250885.2 | XP_006250947.1 | 13 - 14 | 1319 | 105 | ||
XM_006250886.2 | XP_006250948.1 | 14 - 15 | 1431 | 105 | ||
XM_006250887.2 | XP_006250949.1 | 12 - 13 | 1248 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |