Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017361.1 | NP_059057.1 | 4 - 5 | 590 | 153 | |
GenBank mRNA | BC078858.1 | - | 4 - 5 | 620 | 7134682 | 153 |
U63840.1 | - | 4 - 5 | 590 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |