Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107963.1 | NP_001101433.1 | 4 - 5 | 795 | 91 | |
XM_008764009.1 | XP_008762231.1 | 4 - 5 | 717 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |