Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030828.1 | NP_110455.1 | 5 - 6 | 1236 | 98 | |
GenBank mRNA | BC061572.1 | - | 5 - 6 | 1234 | 6887032 | 98 |
L02896.1 | - | 5 - 6 | 1236 | 98 | ||
L34067.1 | - | 5 - 6 | 1069 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |